Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053656.2 | NP_446108.2 | 9 | 950 | ||
XM_006249515.2 | XP_006249577.1 | 9 | 1143 | |||
XM_006249516.2 | XP_006249578.1 | 9 | 1142 | |||
XM_006249517.2 | XP_006249579.1 | 9 | 1105 | |||
XM_008769305.1 | XP_008767527.1 | 8 | 1026 | |||
XM_008769306.1 | XP_008767528.1 | 9 | 819 | |||
XM_008769307.1 | XP_008767529.1 | 7 | 891 | |||
GenBank mRNA | AF013241.1 | - | 2 | 175 | ||
AF020756.1 | - | 8 | 850 | |||
AF020757.1 | - | 8 | 896 | |||
AF020758.1 | - | 7 | 990 | |||
AF020759.1 | - | 9 | 912 | |||
AF064549.1 | - | 8 | 987 | |||
U14414.1 | - | 9 | 950 | |||
Y10473.1 | - | 9 | 950 | |||
Y10474.1 | - | 9 | 932 | |||
Y10475.1 | - | 9 | 986 |