Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053656.2 | NP_446108.2 | 7 | 674 | ||
XM_006249515.2 | XP_006249577.1 | 7 | 867 | |||
XM_006249516.2 | XP_006249578.1 | 7 | 866 | |||
XM_006249517.2 | XP_006249579.1 | 7 | 829 | |||
XM_008769305.1 | XP_008767527.1 | 6 | 750 | |||
XM_008769306.1 | XP_008767528.1 | 7 | 543 | |||
XM_008769307.1 | XP_008767529.1 | 5 | 615 | |||
GenBank mRNA | AF020756.1 | - | 6 | 574 | ||
AF020757.1 | - | 6 | 620 | |||
AF020758.1 | - | 5 | 714 | |||
AF020759.1 | - | 7 | 636 | |||
AF064549.1 | - | 6 | 711 | |||
U14414.1 | - | 7 | 674 | |||
Y10473.1 | - | 7 | 674 | |||
Y10474.1 | - | 7 | 656 | |||
Y10475.1 | - | 7 | 710 |