Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015891.1 | NP_001015891.1 | 2 | 384 | ||
NM_016283.4 | NP_057367.1 | 2 | 147 | |||
GenBank mRNA | AF110777.1 | - | 3 | 396 | ||
AF151895.1 | - | 2 | 87 | |||
AJ878880.1 | - | 2 | 76 | |||
AJ878881.1 | - | 2 | 76 | |||
BC007426.2 | - | 2 | 72 | 2966411 | ||
BX117839.1 | - | 2 | 384 |
Gene Symbol: TAF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015892.1 | NP_001015892.1 | 2 | 384 | ||
NM_003187.4 | NP_003178.1 | 2 | 147 | |||
GenBank mRNA | AI879232.1 | - | 2 | 104 | ||
BC003400.1 | - | 2 | 75 | 3448836 | ||
BC007349.2 | - | 2 | 72 | 2966411 | ||
BC033320.1 | - | 2 | 935 | 4837792 | ||
BG490028.1 | - | 2 | 207 | |||
BG720149.1 | - | 2 | 127 | |||
CX784292.1 | - | 2 | 186 | |||
U21858.1 | - | 2 | 64 |