Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yme1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053682.2 | NP_446134.2 | 15 | 1781 | ||
XM_006254339.2 | XP_006254401.1 | 15 | 1802 | |||
GenBank mRNA | AF151784.1 | - | 15 | 1645 | ||
BC081751.1 | - | 15 | 1781 | 7108995 |
Gene Symbol: LOC100910779 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751732.3 | XP_003751780.1 | 2019 | |||
XM_006254369.2 | XP_006254431.1 | 1802 |