Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC299312 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773045.1 | XP_008771267.1 | 1 | 1184 |
Gene Symbol: Gtpbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771688.1 | XP_008769910.1 | 11 | 1240 | ||
GenBank mRNA | AF325355.1 | - | 11 | 1165 |