Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191861.1 | NP_001178790.1 | 2, 3 | 377 | ||
XM_006236019.2 | XP_006236081.1 | 3, 4 | 841 | |||
XM_006236020.2 | XP_006236082.1 | 3, 4 | 775 | |||
XM_006236021.1 | XP_006236083.1 | 3, 4 | 717 | |||
XM_008762679.1 | XP_008760901.1 | 3, 4 | 692 | |||
GenBank mRNA | AF352168.1 | - | 2, 3 | 305 |