Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053696.1 | NP_446148.1 | 3 | 225 | ||
XM_006256694.1 | XP_006256756.1 | 3 | 247 | |||
XM_006256695.1 | XP_006256757.1 | 3 | 247 | |||
XM_006256696.1 | XP_006256758.1 | 3 | 247 | |||
GenBank mRNA | AF355190.1 | - | 3 | 189 | ||
BC087677.1 | - | 3 | 182 | 7318884 | ||
DN933310.1 | - | 2 | 90 | |||
DY319481.1 | - | 3 | 225 | |||
FQ210873.1 | - | 3 | 204 | |||
FQ220834.1 | - | 3 | 200 | |||
FQ231739.1 | - | 3 | 200 | |||
FQ232482.1 | - | 3 | 203 | |||
FQ235187.1 | - | 202 |