Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005645.3 | NP_005636.1 | 4 | 320 | ||
GenBank mRNA | AK312158.1 | - | 4 | 316 | ||
AU280379.1 | - | 4 | 320 | |||
AV713996.1 | - | 4 | 267 | |||
BC121180.1 | - | 4 | 294 | 40123243 | ||
BC121181.1 | - | 4 | 294 | 40123247 | ||
BG675152.1 | - | 2 | 128 | |||
X84003.1 | - | 4 | 276 |