Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053707.2 | NP_446159.1 | 4 | 689 | ||
GenBank mRNA | AF389348.1 | - | 4 | 418 | ||
AF448810.1 | - | 4 | 418 | |||
BC070943.1 | - | 4 | 689 | 7103046 | ||
FQ231256.1 | - | 5 | 452 |