Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053718.2 | NP_446170.1 | 4 | 959 | ||
GenBank mRNA | AJ006295.1 | - | 4 | 959 | ||
BC129089.1 | - | 663 |