Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aatf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053720.1 | NP_446172.1 | 5 | 751 | ||
GenBank mRNA | AJ238717.3 | - | 5 | 751 | ||
BC078769.1 | - | 5 | 728 | 7110171 |