Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053726.2 | NP_446178.2 | 1, 2 | 216 | ||
XM_006228873.2 | XP_006228935.1 | 1, 2 | 294 | |||
XM_006228874.2 | XP_006228936.1 | 1, 2 | 294 | |||
GenBank mRNA | AB126813.1 | - | 1, 2 | 216 | ||
BC127467.1 | - | 1, 2 | 187 | 7313174 |