Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053736.2 | NP_446188.2 | 7 | 1089 | ||
GenBank mRNA | AY029283.1 | - | 7 | 1112 | ||
BC104720.1 | - | 7 | 1089 | 7382967 | ||
FQ227402.1 | - | 7 | 1126 | |||
FQ232279.1 | - | 7 | 1126 |