Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053738.1 | NP_446190.1 | 5 | 887 | ||
XM_006241395.2 | XP_006241457.1 | 5 | 902 | |||
GenBank mRNA | AY030278.1 | - | 5 | 887 | ||
BC072473.1 | - | 5 | 713 | 7100360 |