Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Becn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034117.1 | NP_001029289.1 | 6 | 626 | ||
NM_053739.2 | NP_446191.1 | 6 | 648 | |||
GenBank mRNA | AY033824.1 | - | 5 | 424 | ||
BC074011.1 | - | 6 | 513 | 7100185 | ||
CK600131.1 | - | 6 | 626 |