Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trafd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053760.2 | NP_446212.2 | 8 | 1056 | ||
XM_006249376.2 | XP_006249438.1 | 8 | 1112 | |||
XM_008769209.1 | XP_008767431.1 | 8 | 1159 | |||
GenBank mRNA | AF329825.1 | - | 2 | 60 |