Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zyx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053761.1 | NP_446213.1 | 9 | 1751 | ||
XM_006236387.2 | XP_006236449.1 | 9 | 1906 | |||
XM_006236388.2 | XP_006236450.1 | 9 | 1869 | |||
XM_006236389.2 | XP_006236451.1 | 8 | 1813 | |||
GenBank mRNA | AF329827.1 | - | 2 | 195 | ||
FQ234073.1 | - | 3 | 348 |