Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025624.2 | NP_001020795.1 | 5 | 356 | ||
GenBank mRNA | BC099178.1 | - | 5 | 322 | 7383003 | |
BC099815.1 | - | 5 | 331 | 7929654 | ||
CB614313.1 | - | 5 | 365 | |||
FQ213300.1 | - | 5 | 324 | |||
FQ213727.1 | - | 5 | 315 | |||
FQ214234.1 | - | 3 | 151 | |||
FQ214394.1 | - | 5 | 315 | |||
FQ214422.1 | - | 5 | 326 | |||
FQ215102.1 | - | 5 | 332 | |||
FQ216489.1 | - | 5 | 287 | |||
FQ217912.1 | - | 5 | 326 | |||
FQ219745.1 | - | 5 | 391 | |||
FQ219836.1 | - | 5 | 353 | |||
FQ220601.1 | - | 5 | 367 | |||
FQ221300.1 | - | 5 | 297 | |||
FQ223249.1 | - | 5 | 315 | |||
FQ229179.1 | - | 5 | 315 | |||
FQ229184.1 | - | 5 | 289 |