Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153814.2 | NP_722521.1 | 9 | 2389 | ||
XM_008767532.1 | XP_008765754.1 | 8 | 6768 | |||
XM_008767533.1 | XP_008765755.1 | 8 | 6768 | |||
XM_008767534.1 | XP_008765756.1 | 8 | 6768 | |||
XM_008767535.1 | XP_008765757.1 | 8 | 6768 | |||
XM_008767536.1 | XP_008765758.1 | 7 | 1987 | |||
GenBank mRNA | AF290213.2 | - | 9 | 2392 | ||
EF116288.1 | - | 1 | 279 |