Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053772.2 | NP_446224.1 | 4 | 443 | ||
XM_006232153.2 | XP_006232215.1 | 2 | 639 | |||
XM_006232155.2 | XP_006232217.1 | 3 | 290 | |||
XM_006232156.2 | XP_006232218.1 | 3 | 2342 | |||
XM_008760857.1 | XP_008759079.1 | 3 | 767 | |||
GenBank mRNA | FM055544.1 | - | 4 | 448 | ||
FQ211701.1 | - | 3 | 350 | |||
L02615.1 | - | 2 | 126 |