Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004178.4 | NP_004169.3 | 2 | 136 | ||
NM_134323.1 | NP_599150.1 | 2 | 601 | |||
NM_134324.2 | NP_599151.2 | 2 | 176 | |||
XM_005269114.1 | XP_005269171.1 | 2 | 171 | |||
XM_005269115.2 | XP_005269172.1 | 2 | 178 | |||
XM_005269117.1 | XP_005269174.1 | 2 | 220 | |||
XM_005269120.4 | XP_005269177.1 | 2 | 601 | |||
XM_005269122.3 | XP_005269179.1 | 2 | 173 | |||
XM_011538712.2 | XP_011537014.1 | 2 | 159 | |||
XM_017019910.1 | XP_016875399.1 | 2 | 171 | |||
XM_017019911.1 | XP_016875400.1 | 2 | 231 | |||
GenBank mRNA | AK290737.1 | - | 2 | 174 | ||
AK293728.1 | - | 2 | 172 | |||
AK309569.1 | - | 2 | 220 | |||
BC002419.2 | - | 2 | 601 | |||
BC005860.2 | - | 2 | 466 | 2960701 | ||
BI771121.1 | - | 2 | 179 | |||
BT007140.1 | - | 2 | 113 | |||
M60801.1 | - | 2 | 104 | |||
U08998.2 | - | 2 | 201 |