Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053788.2 | NP_446240.2 | 9 | 725 | ||
GenBank mRNA | AF217191.1 | - | 9 | 694 | ||
D10392.1 | - | 9 | 725 | |||
D12519.1 | - | 9 | 685 | |||
M95734.1 | - | 9 | 686 |