Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053792.1 | NP_446244.1 | 15 | 1527 | ||
GenBank mRNA | AF226993.1 | - | 15 | 1527 |