Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287742.1 | NP_001274671.1 | 10 | 2063 | ||
NM_001287743.1 | NP_001274672.1 | 9 | 1865 | |||
NM_001287744.1 | NP_001274673.1 | 8 | 1692 | |||
NM_001287745.1 | NP_001274674.1 | 9 | 1771 | |||
NM_001287747.1 | NP_001274676.1 | 8 | 1728 | |||
NM_001287748.1 | NP_001274677.1 | 8 | 1390 | |||
NM_001287749.1 | NP_001274678.1 | 8 | 1628 | |||
NM_001287750.1 | NP_001274679.1 | 7 | 1630 | |||
NM_001287751.1 | NP_001274680.1 | 7 | 1502 | |||
NM_001287756.1 | NP_001274685.1 | 5 | 1087 | |||
NM_004462.4 | NP_004453.3 | 8 | 1574 | |||
XM_017013217.1 | XP_016868706.1 | 10 | 2355 | |||
GenBank mRNA | AK057726.1 | - | 8 | 1692 | ||
AK098682.1 | - | 1390 | ||||
AK293545.1 | - | 8 | 1628 | |||
AK296043.1 | - | 7 | 1317 | |||
AK300059.1 | - | 7 | 1392 | |||
AK301617.1 | - | 5 | 948 | |||
AK315993.1 | - | 9 | 1641 | |||
AK316033.1 | - | 8 | 1595 | |||
AK316182.1 | - | 7 | 1590 | |||
AK316351.1 | - | 8 | 1355 | |||
AK316531.1 | - | 8 | 1340 | |||
AK316534.1 | - | 7 | 1502 | |||
BC009251.2 | - | 8 | 1421 | 2961623 | ||
BC029641.1 | - | 8 | 1384 | 4514761 | ||
BC034440.1 | - | 8 | 1384 | |||
L06105.1 | - | 8 | 1396 | |||
S76822.1 | - | 9 | 1395 | |||
X69141.1 | - | 8 | 1443 |