Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053825.2 | NP_446277.1 | 6, 7 | 837 | ||
XM_006254229.2 | XP_006254291.1 | 6, 7 | 891 | |||
XM_006254230.2 | XP_006254292.1 | 6, 7 | 892 | |||
XM_006254231.2 | XP_006254293.1 | 6, 7 | 803 | |||
GenBank mRNA | BC059158.1 | - | 6, 7 | 841 | 6920493 | |
FQ212577.1 | - | 6, 7 | 825 | |||
FQ212651.1 | - | 6, 7 | 835 | |||
FQ214998.1 | - | 6, 7 | 836 | |||
FQ215205.1 | - | 6, 7 | 835 | |||
FQ215286.1 | - | 6, 7 | 810 | |||
FQ219484.1 | - | 6, 7 | 835 | |||
L19927.1 | - | 4, 5 | 715 |