Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053855.1 | NP_446307.1 | 12 | 1534 | ||
XM_006241047.2 | XP_006241109.1 | 12 | 1756 | |||
GenBank mRNA | BC166580.1 | - | 12 | 1534 | 7380301 | |
U01914.1 | - | 12 | 1509 |