Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cux1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751160.3 | XP_003751208.1 | 16, 17 | 2285 | ||
XM_003751161.3 | XP_003751209.1 | 16, 17 | 1978 | |||
XM_006249202.2 | XP_006249264.1 | 16, 17 | 2985 | |||
XM_006249204.2 | XP_006249266.1 | 16, 17 | 2684 | |||
XM_006249205.2 | XP_006249267.1 | 16, 17 | 2678 | |||
XM_347163.7 | XP_347164.4 | 16, 17 | 2991 |