Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlgn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053868.2 | NP_446320.1 | 7 | 2384 | ||
XM_008760890.1 | XP_008759112.1 | 4 | 1355 | |||
GenBank mRNA | U22952.1 | - | 7 | 2748 |