Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053879.1 | NP_446331.1 | 12 | 1529 | ||
XM_008763166.1 | XP_008761388.1 | 8 | 1260 | |||
GenBank mRNA | U35371.1 | - | 12 | 1529 |