Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1i2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270624.1 | NP_001257553.1 | 13 | 1606 | ||
NM_001270625.1 | NP_001257554.1 | 12 | 1546 | |||
NM_053880.2 | NP_446332.2 | 14 | 1624 | |||
XM_006234344.2 | XP_006234406.1 | 13 | 1383 | |||
XM_006234346.2 | XP_006234408.1 | 12 | 1322 | |||
GenBank mRNA | BC078764.1 | - | 13 | 1352 | 7113840 | |
U39044.1 | - | 14 | 1340 | |||
U39045.1 | - | 13 | 1322 | |||
U39046.1 | - | 12 | 1262 |