Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053881.1 | NP_446333.1 | 2, 3 | 176 | ||
XM_006245134.2 | XP_006245196.1 | 2, 3 | 244 | |||
GenBank mRNA | FQ211652.1 | - | 2, 3 | 235 | ||
U40652.1 | - | 2, 3 | 142 | |||
X92563.1 | - | 2, 3 | 176 |