Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBL1X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139466.1 | NP_001132938.1 | 16 | 1891 | ||
NM_001139467.1 | NP_001132939.1 | 15 | 1746 | |||
NM_001139468.1 | NP_001132940.1 | 16 | 1881 | |||
NM_005647.3 | NP_005638.1 | 16 | 2010 | |||
XM_011545571.2 | XP_011543873.1 | 15 | 1732 | |||
GenBank mRNA | AB209608.1 | - | 11 | 1964 | ||
AK289409.1 | - | 15 | 1746 | |||
AK290962.1 | - | 16 | 1874 | |||
BC032708.1 | - | 16 | 1857 | 5496758 | ||
BC052304.1 | - | 16 | 2010 | 6378852 | ||
Y12781.1 | - | 16 | 2169 |