Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172085.1 | NP_001165556.1 | 2 | 208 | ||
NM_003194.4 | NP_003185.1 | 3 | 410 | |||
GenBank mRNA | AK298272.1 | - | 3 | 361 | ||
AK304648.1 | - | 2 | 198 | |||
AK315222.1 | - | 3 | 376 | |||
BC109053.1 | - | 3 | 324 | 40032492 | ||
BC109054.1 | - | 3 | 328 | 40032493 | ||
BC110341.1 | - | 3 | 372 | 5761426 | ||
BM720195.1 | - | 3 | 410 | |||
BT019657.1 | - | 2 | 131 | |||
CR456776.1 | - | 2 | 131 | |||
FJ770048.1 | - | 2 | 212 | |||
M34960.1 | - | 2 | 131 | |||
M55654.1 | - | 3 | 372 | |||
X54993.1 | - | 2 | 229 | |||
Z22828.1 | - | 3 | 369 |