Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172085.1 | NP_001165556.1 | 3 | 597 | ||
NM_003194.4 | NP_003185.1 | 4 | 799 | |||
GenBank mRNA | AK298272.1 | - | 5 | 681 | ||
AK304648.1 | - | 3 | 584 | |||
AK315222.1 | - | 4 | 762 | |||
BC109053.1 | - | 4 | 710 | 40032492 | ||
BC109054.1 | - | 5 | 564 | 40032493 | ||
BC110341.1 | - | 4 | 758 | 5761426 | ||
BT019657.1 | - | 3 | 511 | |||
CR456776.1 | - | 3 | 520 | |||
M34960.1 | - | 3 | 520 | |||
M55654.1 | - | 4 | 761 | |||
X54993.1 | - | 3 | 606 | |||
Z22828.1 | - | 4 | 758 |