Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NFASC SIRNA Assays ›
Gene Symbol:
Nfasc
Gene Name:
neurofascin
Gene Aliases:
NF
Chromosome Location:
Chr.13: 49335407 - 49522470 on Build Rnor_6.0
UniGene ID:
Rn.3048
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s138288
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nfasc
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001160313.1
NP_001153785.1
2
260
NM_001160314.1
NP_001153786.1
5
496
NM_001160315.1
NP_001153787.1
4
687
NM_053909.2
NP_446361.1
4
478
XM_006249727.2
XP_006249789.1
5
551
XM_006249734.2
XP_006249796.1
5
550
XM_006249738.2
XP_006249800.1
4
531
XM_006249741.2
XP_006249803.1
5
472
XM_006249742.2
XP_006249804.1
5
421
XM_008769406.1
XP_008767628.1
5
676
XM_008769407.1
XP_008767629.1
4
658
XM_008769408.1
XP_008767630.1
5
676
XM_008769409.1
XP_008767631.1
5
676
XM_008769410.1
XP_008767632.1
4
658
XM_008769411.1
XP_008767633.1
5
676
XM_008769412.1
XP_008767634.1
5
676
XM_008769413.1
XP_008767635.1
5
676
XM_008769414.1
XP_008767636.1
5
675
XM_008769415.1
XP_008767637.1
5
675
XM_008769416.1
XP_008767638.1
4
657
XM_008769417.1
XP_008767639.1
4
657
XM_008769418.1
XP_008767640.1
4
657
XM_008769419.1
XP_008767641.1
4
657
GenBank mRNA
AY061639.1
-
2
260
L11002.1
-
1227
U81035.1
-
2
98
U81036.1
-
3
116
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001160318, NM_182716, NM_001160316, NM_001160317, AK129207, and others.
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein localization to paranode region of axon
cell adhesion
axon guidance
peripheral nervous system development
transmission of nerve impulse
paranodal junction assembly
heterotypic cell-cell adhesion
myelination
clustering of voltage-gated sodium channels
synapse organization
protein localization to juxtaparanode region of axon
protein targeting to plasma membrane
protein domain specific binding
protein binding involved in heterotypic cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-gx66z:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline