Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105748.1 | NP_001099218.1 | 1 | 696 | ||
GenBank mRNA | U89873.1 | - | 1 | 241 |