Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053917.1 | NP_446369.1 | 11 | 1115 | ||
XM_008772462.1 | XP_008770684.1 | 11 | 958 | |||
XM_008772463.1 | XP_008770685.1 | 11 | 958 | |||
XM_008772464.1 | XP_008770686.1 | 11 | 1115 | |||
XM_008772465.1 | XP_008770687.1 | 10 | 907 | |||
GenBank mRNA | U96920.1 | - | 11 | 1115 | ||
U96921.1 | - | 11 | 1115 |