Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RIMS2 SIRNA Assays ›
Gene Symbol:
Rims2
LOC103692905
Gene Name:
regulating synaptic membrane exocytosis 2
regulating synaptic membrane exocytosis protein 2-like
Gene Aliases:
Nim2, Rim2
-
Chromosome Location:
Chr.7: 78092020 - 78594162 on Build Rnor_6.0
Chromosome Location:
Chr.7: 78671923 - 78677321 on Build Rnor_6.0
UniGene ID:
Rn.161948
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID
s138402
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rims2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053945.2
NP_446397.1
27, 28
4369
NM_145881.2
NP_665888.1
4, 5
691
XM_006241591.2
XP_006241653.1
25, 26
4437
XM_006241592.2
XP_006241654.1
24, 25
4371
XM_006241593.2
XP_006241655.1
26, 27
4337
XM_006241594.2
XP_006241656.1
25, 26
4295
XM_006241598.2
XP_006241660.1
25, 26
7141
XM_008765437.1
XP_008763659.1
28, 29
6639
XM_008765438.1
XP_008763660.1
26, 27
4488
XM_008765439.1
XP_008763661.1
24, 25
6333
XM_008765440.1
XP_008763662.1
22, 23
3918
XM_008765441.1
XP_008763663.1
23, 24
3825
XM_008765442.1
XP_008763664.1
22, 23
3777
XM_008765443.1
XP_008763665.1
21, 22
3710
XM_008765445.1
XP_008763667.1
5, 6
1619
XM_008765446.1
XP_008763668.1
5, 6
3552
XM_008765447.1
XP_008763669.1
25, 26
4311
GenBank mRNA
AF199322.1
-
27, 28
4369
AF199323.1
-
22, 23
3760
AF199324.1
-
22, 23
3901
AF199325.1
-
23, 24
3808
AF199326.1
-
22, 23
3760
AF199327.1
-
22, 23
3760
AF199328.1
-
21, 22
3580
AF199329.1
-
21, 22
3694
AF199330.1
-
26, 27
4321
AF199331.1
-
25, 26
4279
AF199332.1
-
22, 23
3760
AF199335.1
-
4, 5
692
AF548738.1
-
25, 26
3792
Gene Symbol: LOC103692905
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_008765484.1
XP_008763706.1
262
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
exocytosis
positive regulation of gene expression
calcium ion regulated exocytosis
regulation of exocytosis
cAMP-mediated signaling
insulin secretion
regulation of membrane potential
calcium ion-regulated exocytosis of neurotransmitter
positive regulation of inhibitory postsynaptic potential
positive regulation of excitatory postsynaptic potential
protein binding
Rab GTPase binding
protein domain specific binding
ion channel binding
metal ion binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0