Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053950.1 | NP_446402.1 | 8 | 768 | ||
GenBank mRNA | FQ233756.1 | - | 3 | 240 | ||
Z48225.1 | - | 8 | 768 |