Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053970.2 | NP_446422.2 | 5, 6 | 694 | ||
XM_006231880.2 | XP_006231942.1 | 5, 6 | 848 | |||
XM_006231881.2 | XP_006231943.1 | 6, 7 | 926 | |||
GenBank mRNA | BC072687.1 | - | 5, 6 | 694 | 7099208 | |
X87157.1 | - | 5, 6 | 802 |