Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053971.3 | NP_446423.2 | 5, 6 | 751 | ||
XM_006249377.1 | XP_006249439.1 | 6, 7 | 770 | |||
XM_006249378.2 | XP_006249440.1 | 6, 7 | 804 | |||
GenBank mRNA | BC061784.1 | - | 5, 6 | 747 | ||
BC078761.1 | - | 5, 6 | 754 | 7123086 | ||
BC097295.1 | - | 5, 6 | 755 | 7446680 | ||
FQ213055.1 | - | 4, 5 | 429 | |||
FQ216758.1 | - | 4, 5 | 429 | |||
FQ217744.1 | - | 4, 5 | 429 | |||
FQ219956.1 | - | 4, 5 | 429 | |||
FQ219982.1 | - | 4, 5 | 429 | |||
FQ220180.1 | - | 4, 5 | 429 | |||
FQ220346.1 | - | 4, 5 | 429 | |||
FQ220362.1 | - | 4, 5 | 429 | |||
FQ220471.1 | - | 4, 5 | 429 | |||
FQ220678.1 | - | 4, 5 | 429 | |||
FQ220699.1 | - | 4, 5 | 429 | |||
FQ220887.1 | - | 4, 5 | 429 | |||
FQ220959.1 | - | 4, 5 | 429 | |||
FQ221615.1 | - | 4, 5 | 429 | |||
FQ222331.1 | - | 4, 5 | 429 | |||
FQ222465.1 | - | 4, 5 | 429 | |||
FQ222658.1 | - | 4, 5 | 429 | |||
FQ223301.1 | - | 4, 5 | 429 | |||
FQ227060.1 | - | 4, 5 | 429 | |||
FQ227100.1 | - | 4, 5 | 429 | |||
FQ227955.1 | - | 4, 5 | 429 | |||
FQ227974.1 | - | 4, 5 | 429 | |||
FQ228583.1 | - | 4, 5 | 429 | |||
FQ228665.1 | - | 4, 5 | 429 | |||
FQ229345.1 | - | 4, 5 | 429 | |||
FQ229355.1 | - | 4, 5 | 429 | |||
FQ229484.1 | - | 6, 7 | 769 | |||
FQ229571.1 | - | 4, 5 | 429 | |||
FQ229624.1 | - | 4, 5 | 429 | |||
FQ229675.1 | - | 4, 5 | 429 | |||
FQ229700.1 | - | 4, 5 | 429 | |||
FQ229820.1 | - | 4, 5 | 429 | |||
FQ229854.1 | - | 4, 5 | 429 | |||
FQ229857.1 | - | 4, 5 | 428 | |||
FQ229943.1 | - | 6, 7 | 768 | |||
FQ230171.1 | - | 4, 5 | 429 | |||
FQ230222.1 | - | 4, 5 | 429 | |||
FQ232778.1 | - | 4, 5 | 429 | |||
FQ233198.1 | - | 4, 5 | 429 | |||
FQ233475.1 | - | 4, 5 | 429 | |||
FQ235095.1 | - | 4, 5 | 429 | |||
X87107.1 | - | 6, 7 | 765 |