Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053997.4 | NP_446449.2 | 2 | 1853 | ||
XM_006228965.2 | XP_006229027.1 | 2 | 2047 | |||
XM_006228967.2 | XP_006229029.1 | 2 | 2047 | |||
XM_006228971.2 | XP_006229033.1 | 2 | 2047 | |||
XM_006228972.2 | XP_006229034.1 | 2 | 2047 | |||
XM_006228973.2 | XP_006229035.1 | 2 | 2047 | |||
XM_008759306.1 | XP_008757528.1 | 2 | 2047 | |||
XM_008759307.1 | XP_008757529.1 | 2 | 2047 | |||
XM_008759308.1 | XP_008757530.1 | 2 | 2047 | |||
XM_008759309.1 | XP_008757531.1 | 2 | 2047 | |||
GenBank mRNA | AY179603.1 | - | 2 | 1853 | ||
AY179604.1 | - | 2 | 1853 | |||
AY822673.1 | - | 2 | 1853 | |||
AY822674.1 | - | 2 | 1853 | |||
M84210.1 | - | 2 | 1853 | |||
M84211.2 | - | 2 | 1853 |