Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003198.2 | NP_003189.2 | 6 | 1802 | ||
GenBank mRNA | AF116729.1 | - | 6 | 1651 | ||
AK310229.1 | - | 6 | 1788 | |||
AK313079.1 | - | 6 | 1674 | |||
BC002883.2 | - | 6 | 1686 | 3941728 | ||
BC020448.1 | - | 3 | 414 | |||
L47345.1 | - | 6 | 1696 |