Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005139.2 | NP_005130.1 | 11, 12 | 1093 | ||
GenBank mRNA | AK313945.1 | - | 11, 12 | 868 | ||
BC000871.1 | - | 11, 12 | 925 | 3460409 | ||
CR407648.1 | - | 9, 10 | 783 | |||
M20560.1 | - | 10, 11 | 829 | |||
M63310.1 | - | 10, 11 | 819 |