Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054006.1 | NP_446458.1 | 12, 13 | 1583 | ||
GenBank mRNA | FQ222050.1 | - | 9, 10 | 1182 | ||
FQ223276.1 | - | 11, 12 | 1488 | |||
X52311.1 | - | 12, 13 | 1583 |
Gene Symbol: LOC100364335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751853.3 | XP_003751901.1 | 1 | 1584 |