Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tenm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020088.1 | NP_064473.1 | 19, 20 | 3922 | ||
GenBank mRNA | AF086607.1 | - | 19, 20 | 3922 |