Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005996.3 | NP_005987.3 | 2 | 1601 | ||
NM_016569.3 | NP_057653.3 | 2 | 1601 | |||
GenBank mRNA | AF002228.1 | - | 2 | 637 | ||
AF170708.2 | - | 2 | 897 | |||
AF216750.1 | - | 2 | 753 | |||
AK054604.1 | - | 2 | 1604 | |||
AK304639.1 | - | 2 | 871 | |||
AY034105.1 | - | 1 | 84 | |||
BC025258.1 | - | 2 | 1027 | 5458526 |