Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057123.1 | NP_476464.1 | 8 | 1015 | ||
GenBank mRNA | BC063157.1 | - | 9 | 1027 | 6919865 | |
D50696.1 | - | 8 | 1015 | |||
FQ209916.1 | - | 9 | 1020 |