Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000545.6 | NP_000536.5 | 7 | 1540 | ||
NM_001306179.1 | NP_001293108.1 | 7 | 1540 | |||
XM_005253931.3 | XP_005253988.1 | 7 | 1593 | |||
GenBank mRNA | BC104908.1 | - | 7 | 1359 | 8143911 | |
BC104910.1 | - | 7 | 1359 | 8143913 | ||
BC143483.1 | - | 7 | 1360 | |||
HM116541.1 | - | 7 | 1482 | |||
HM116542.1 | - | 7 | 1156 | |||
HM116543.1 | - | 5 | 945 | |||
HM116545.1 | - | 4 | 469 | |||
HM116546.1 | - | 2 | 99 | |||
HM116547.1 | - | 7 | 1482 | |||
HM116557.1 | - | 6 | 1270 | |||
HM116558.1 | - | 6 | 1215 | |||
HM116559.1 | - | 3 | 410 | |||
HM116560.1 | - | 4 | 334 | |||
HM116561.1 | - | 2 | 203 | |||
HM116562.1 | - | 2 | 55 | |||
HM116563.1 | - | 3 | 346 | |||
HM449088.1 | - | 7 | 1362 | |||
HM449089.1 | - | 6 | 1162 | |||
M57732.1 | - | 7 | 1362 | |||
X71346.1 | - | 7 | 1535 | |||
X71347.1 | - | 6 | 2206 |