Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khdrbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130405.1 | NP_569089.1 | 3 | 613 | ||
GenBank mRNA | AF305619.1 | - | 3 | 613 | ||
AF393783.1 | - | 3 | 530 | |||
BC061987.1 | - | 3 | 637 | 5599491 |